missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Rat HO-1/HMOX1/HSP32 His Protein

Product Code. 18348604 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
200 μg
50 μg
Unit Size:
100µg
200µg
50µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18348604 200 μg 200µg
18345734 50 μg 50µg
18331364 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18348604 Supplier Novus Biologicals™ Supplier No. NBP318328200UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) SDS-PAGE, Western Blot
Formulation 50mM Tris/HCl pH7.5, 5mM Bme, 0.15NaCl, 10% glycerol
Gene ID (Entrez) 3162
Molecular Weight (g/mol) M.W. Theoretical: 32 kDa
Quantity 200 μg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias bK286B10, EC 1.14.99.3, heat shock protein, 32-kD, heme oxygenase (decycling) 1, HO, HO1, HO-1heme oxygenase 1, HSP32
Gene Symbol HMOX1
Research Category Apoptosis, Cellular Markers, ER Markers, Lipid and Metabolism
Purity or Quality Grade >85%
Protein HO-1/HMOX1/HSP32
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.