missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Rat gp130 Fc Chimera Protein
Description
The Recombinant Rat gp130 Fc Chimera Protein is derived from NS0. The Recombinant Rat gp130 Fc Chimera Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 25205 |
| Molecular Weight (g/mol) | 94 kDa (monomer) |
| Name | gp130 Fc Chimera |
| Quantity | 100μg |
| Source | Mouse myeloma cell line, NS0-derived rat gp130 protein Rat gp130 (Gln23 - Glu618) (C-terminus) Accession # P15725 IEGRMDP Mouse IgG2a (Glu98 - Lys330) (N-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, ≤ -20 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, ≤ -20 °C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?