missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat GMF B Protein

A cDNA sequence encoding the GMF B was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12002-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15959058

  • £5160.00 / 1mg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

GMF B Protein
Research Use Only
Recombinant Protein
E. coli
SESLVVCDVA EDLVEKLRKF RFRKETHNAA IIMKIDKDKR LVVLDEELEG VSPDELKDEL PERQPRFIVY SYKYQHDDGR VSYPLCFIFS SPLGCKPEQQ MMYAGSKNKL VQTAELTKVF EIRNTEDLTE EWLREKLGFF H
Greater than 97.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Rat
Untagged
Lyophilized from a 0.2microm filtered concentrated (1.0 mg/ml) solution in 1M PBS, pH 7.4.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers