missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat GM-CSF / CSF2 Protein
Recombinant Protein
£80.80 - £3195.00
Specifications
Accession Number | NP_446304.1 |
---|---|
Gene ID (Entrez) | 116630 |
Name | GM-CSF / CSF2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15976249
|
enQuireBio™
QP5495-5UG |
5 μg |
£80.80
5µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956249
|
enQuireBio™
QP5495-20UG |
20 μg |
£191.00
20µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15936249
|
enQuireBio™
QP5495-100UG |
100 μg |
£476.00
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966249
|
enQuireBio™
QP5495-500UG |
500 μg |
£1965.00
500µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15946249
|
enQuireBio™
QP5495-1MG |
1 mg |
£3195.00
1mg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_446304.1 | |
GM-CSF / CSF2 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 20 pg/mL in the FDCP-1 cell proliferation assay. The bioactivity acceptance criteria is set at 5.0 x 10^7 units/mg. | |
Rat | |
Untagged | |
20 mM sodium bicarbonate buffer, pH 8.5 |
116630 | |
Research Use Only | |
CSF2 | |
Recombinant Protein | |
E. coli | |
MAPTRSPNPV TRPWKHVDAI KEALSLLNDM RALENEKNED VDIISNEFSI QRPTCVQTRL KLYKQGLRGN LTKLNGALTM IASHYQTNCP PTPETDCEIE VTTFEDFIKN LKGFLFDIPF DCWKPVQK | |
Greater than 95% as determined by SDS-PAGE. |