missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Glypican-3 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Glypican-3, was expressed in the hosts and tags indicated.
£223.39 - £2480.00
Specifications
Accession Number | NP_036906.1 |
---|---|
Gene ID (Entrez) | 25236 |
Name | Glypican-3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16027741
|
enQuireBio™
QP9158-YE-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16047741
|
enQuireBio™
QP9158-YE-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16037741
|
enQuireBio™
QP9158-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057741
|
enQuireBio™
QP9158-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067741
|
enQuireBio™
QP9158-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16077741
|
enQuireBio™
QP9158-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_036906.1 | |
Glypican-3 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
25236 | |
Research Use Only | |
GPC3 | |
Recombinant Protein | |
Yeast | |
QPPPPPDATC HQVRSFFQRL QPGLKWVPET PVPGSDLQVC LPKGPTCCSR KMEEKYQLTA RLNMEQLLQS ASMELKFLII QNAAVFQEAF EIVVRHAKNY TNAMFKNNYP SLTPQAFEFV GEFFTDVSLY ILGSDINVDD MVNELFDSLF PVIYTQMMNP GLPESVLDIN ECLRGARRDL KVFGSFPKLI MTQVSKSLQV TRIFLQALNL GIEVINTTDH LKFSKDCGRM LTRMWYCSYC QGLMMVKPCG GYCNVVMQGC MAGVVEIDKY WREYILSLEE LVNGMYRIYD MENVLLGLFS TIHDSIQYVQ KNGGKLTTTI GKLCAHSQQR QYRSAYYPED LFIDKKVLKV ARVEHEETLS SRRRELIQKL KSFISFYSAL PGYICSHSPV AENDTLCWNG QELVERYSQK AARNGMKNQF NLHELKMKGP EPVVSQIIDK LKHINQLLRT MSVPKGKVVD KSLDEEGLES GDCGDDEDEC IGSSGDGMMK VKNQLRFLAE LAYDLDVDDA PGNKQHGNQK DNEITTSHSV GNMPSP | |
Greater than 90% as determined by SDS-PAGE. |