missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GDNF protein is fully biologically active when compared to standard. Measured by its ability to support the survival and stimulate neurite outgrowth of dissociated chick embryonic dorsal root ganglia (DRG) neurons. The ED50 for this effect is typically 1 - 3 ng/mL
Specifications
Specifications
| Accession Number | Q07731, Q07731, Q07731 |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 1 x PBS, pH 7.4. |
| Gene ID (Entrez) | 2668 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 29.8 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/μg of GDNF as determined by LAL method. |
| Gene Alias | Astrocyte-derived trophic factor, ATF, ATF1, ATF2, glial cell derived neurotrophic factor, glial cell line derived neurotrophic factor, glial cell line-derived neurotrophic factor, HFB1-GDNF, hGDNF, HSCR3 |
| Gene Symbol | GDNF |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?