missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Rat GDNF Protein

Product Code. 18397227
Change view
Click to view available options
Quantity:
10 μg
50 μg
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS.
Unit Size:
10µg
50µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Formulation unitSize
18397227 50 μg Lyophilized from a 0.2 μm filtered solution in PBS. 50µg
18127670 10 μg Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. 10µg
18177930 50 μg Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. 50µg
18187430 10 μg Lyophilized from a 0.2 μm filtered solution in PBS. 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18397227 Supplier R&D Systems™ Supplier No. 512GF050/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity, Binding Activity

Measured in a cell proliferation assay using SH-SY5Y human neuroblastoma cells. The ED50 for this effect is 2-12 ng/mL in the presence of Recombinant Rat GFR alpha-1/GDNF R alpha-1 Fc Chimera (Catalog # 560-GR ). Measured by its binding ability in a functional ELISA. Immobilized Recombinant Rat GFR alpha-1/GDNF R alpha-1 Fc Chimera (Catalog # 560-GR ) at 1 μg/mL binds Recombinant Rat GDNF with an apparent Kd <4 nM.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q07731
For Use With (Application) Bioactivity, Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 2668
Molecular Weight (g/mol) M.W. (theoretical): 15 kDa (monomer)
Name GDNF
Quantity 50 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived rat GDNF protein Ser78-Ile211
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Symbol Gdnf
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein GDNF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.