missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Rat G-CSF Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
10 μg
100 μg
1mg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
Description
G-CSF protein is fully biologically active when compared to standard. The ED50 determined by a cell proliferation assay using murine NFS-60 cells is less than 0.05 ng/mL, corresponding to a specific activity of >2.0 x 10^7 IU/mg
Specifications
Specifications
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 5 mM Sodium Citrate, pH 4.0. |
| Gene ID (Entrez) | 1440 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 21.5 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/μg of G-CSF as determined by LAL method. |
| Gene Alias | C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3OS, filgrastim, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin |
| Gene Symbol | CSF3 |
| Reconstitution | Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction