missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Rat FGF-21 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
1mg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
Description
FGF-21 protein is fully biologically active when compared to standard. The ED50 as determined by thymidine uptake assay using FGF-receptors transfected BaF3 cells is less than 0.5 μg/mL, corresponding to a specific activity of >2.0 x 10^3 IU/mg in the presence of 5 μg/mL of rMuKlotho-beta and 10 μg/mL of heparin.
Specifications
Specifications
| Accession Number | Q8VI80 |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, 500 mM NaCl, pH 7.4. |
| Gene ID (Entrez) | 26291 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 19.7 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/μg of FGF-21 as determined by LAL method. |
| Gene Alias | FGF-21, fibroblast growth factor 21 |
| Gene Symbol | FGF21 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction