missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat FGF 21 Protein

Product Code. 15934378
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
This item is not returnable. View return policy

Product Code. 15934378

Brand: enQuireBio™ QP106005ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the FGF 21 was constructed and used to recombinantly synthesize the protein.

Specifications

Name FGF 21 Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity The ED50 determined by a cell proliferation assay using murine NIH/3T3 cells is less than 700 ng/ml, corresponding to a specific activity of > 1.4 M 1000 IU/mg in the presence of 5microg/ml of rMuKlotho-beta.
Product Type Recombinant Protein
Cross Reactivity Rat
Species E. coli
Protein Tag Untagged
Sequence AYPISDSSPL LQFGGQVRQR YLYTDDDQDT EAHLEIREDG TVVGTAHRSP ESLLELKALK PGVIQILGVK ASRFLCQQPD GTLYGSPHFD PEACSFRELL LKDGYNVYQS EAHGLPLRLP QKDSQDPATR GPVRFLPMPG LPHEPQEQPG VLPPEPPDVG SSDPLSMVEP LQGRSPSYAS
Buffer Lyophilized from a 0.2microm filtered concentrated solution in 1M PBS, pH 7.4.
Purity or Quality Grade Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.