missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat FGF 1 Protein
A cDNA sequence encoding the FGF 1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10596-1mg
Additional Details : Weight : 0.01000kg
Specifications
FGF 1 Protein | |
Research Use Only | |
The ED50 as determined by the dose-dependent proliferation of mouse BALB/c 3T3 cells, is less than 0.2ng/ml corresponding to a Specific Activity of 5x106IU/mg. | |
Rat | |
Untagged | |
Lyophilized at a concentration of 1 mg/ml in 5mM Na2PO4, pH 7.5 and 50mM NaCl. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MFNLPLGNYK KPKLLYCSNG GHFLRILPDG TVDGTRDRSD QHIQLQLSAE SAGEVYIKGT ETGQYLAMDT EGLLYGSQTP NEECLFLERL EENHYNTYTS KKHAEKNWFV GLKKNGSCKR GPRTHYGQKA ILFLPLPVSS D | |
Greater than 98.0% as determined by SDS-PAGE. |