missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Dipeptidyl aminopeptidase-like protein 1
A DNA sequence encoding the Rattus norvegicus (Rat) Dipeptidyl aminopeptidase-like protein 6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5945-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_074041.1 | |
Dipeptidyl aminopeptidase-like protein 1 | |
Research Use Only | |
DPP6 | |
Recombinant Protein | |
Yeast | |
LTPAEDTSLS QKKKVTVEDL FSEDFKIHDP EAKWISDKEF IYRERKGSVI LRNVETNNST VLIEGKKIES LRAIRYEISP DKEYALFSYN VEPVYQHSHT GYYVLSKIPH GDPQSLDPPE VSNAKLQYAG WGPKGQQLIF IFENNIYYCA HVGKQAIRVV STGKEGVIYN GLSDWLYEEE ILKSHIAHWW SPDGTRLAYA TINDSRVPLM ELPTYTGSVY PTVKPYHYPK AGSENPSISL HVIGLNGPTH DLEMMPPDDP RMREYYITMV KWATSTKVAV TWLNRAQNVS ILTLCDATTG VCTKKHEDES EAWLHRQNEE PVFSKDGRKF FFVRAIPQGG RGKFYHITVS SSQPNSSNDN IQSITSGDWD VTEILTYDEK RNKLYFLSTE DLPRRRHLYS ANTVDDFNRQ CLSCDLVENC TYVSASFSHN MDFFLLKCEG PGVPTVTVHN TTDKRRMFDL EANEQVQKAI YDRQMPKIEY RKIEVEDYSL PMQILKPATF TDTAHYPLLL VVDGTPGSQS VSERFEVTWE TVLVSSHGAV VVKCDGRGSG FQGTKLLHEV RRRLGFLEEK DQMEAVRTML KEQYIDKTRV AVFGKDYGGY LSTYILPAKG ENQGQTFTCG SALSPITDFK LYASAFSERY LGLHGLDNRA YEMTKLAHRV SALEDQQFLI IHATADEKIH FQHTAELITQ LIKGKANYSL QIYPDESHYF HSVALKQHLY RSIIGFFVEC FRIQDKLPTA TAKEDEEED | |
Greater than 90% as determined by SDS-PAGE. |
29272 | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |