missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Cyclophilin F Protein
A cDNA sequence encoding the Cyclophilin F was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Gene ID (Entrez) | 282819 |
---|---|
Name | Cyclophilin F Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | Ppif |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15974288
|
enQuireBio™
QP10573-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954288
|
enQuireBio™
QP10573-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15964288
|
enQuireBio™
QP10573-1MG |
1 mg |
£5730.00
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
282819 | |
Research Use Only | |
Ppif | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSCSDGGAR GANSSSQNPL VYLDVGADGQ PLGRVVLELK ADVVPKTAEN FRALCTGEKG FGYKGSTFHR VIPAFMCQAG DFTNHNGTGG KSIYGSRFPD ENFTLKHVGP GVLSMANAGP NTNGSQFFIC TIKTDWLDGK HVVFGHVKEG MDVVKKIESF GSKSGKTSKK IVITDCGQLS | |
Greater than 90.0% as determined by SDS-PAGE. |
Cyclophilin F Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 1,300 nmol/min/mg, and is defined as the amount of enzyme that cleaves 1nmol of suc-AAFP-PNA per minute at 37C in Tris–HCl pH 8.0 using chymotrypsin. | |
Rat | |
His | |
Cyclophilin F protein solution (1 mg/ml) containing Phosphate Buffered Saline (pH 7.4), 10% glycerol and 1mM DTT. |