missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Rat CXCL7/Thymus Chemokine-1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
£327.21
Specifications
Gene ID (Entrez) | 246358 |
---|---|
Molecular Weight (g/mol) | 6.8 kDa |
Name | CXCL7/Thymus Chemokine-1 |
Quantity | 25μg |
Source | E. coli-derived rat CXCL7/Thymus Chemokine-1 protein Ile46-Ile107 |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18196630
|
R&D Systems™
1116-CK-025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
£327.21
25µg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18116830
|
R&D Systems™
1116-CK-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
£327.21
25µg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Rat CXCL7/Thymus Chemokine-1 Protein is derived from E. coli. The Recombinant Rat CXCL7/Thymus Chemokine-1 Protein has been validated for the following applications: Bioactivity.Specifications
246358 | |
CXCL7/Thymus Chemokine-1 | |
E. coli-derived rat CXCL7/Thymus Chemokine-1 protein Ile46-Ile107 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
6.8 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |