missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Rat CXCL1/GRO alpha/KC/CINC-1 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
Description
CXCL1 Protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using rat neutrophils is in a concentration range of 10-100 ng/mL.
Specifications
Specifications
| Accession Number | P14095, P14095, P14095 |
| For Use With (Application) | SDS-PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered concentrated solution in 20 mM PB, pH 7.4, 150 mM NaCl. |
| Gene ID (Entrez) | 2919 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 7.8 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Endotoxin Concentration | Less than 1 EU/μg of CXCL1/GRO alpha/KC/CINC-1 AS determined by LAL method. |
| Gene Alias | CINC-1/CXCL1, FSP, GRO1, GROa, MGSA, MGSA-a, NAP-3, SCYB1 |
| Gene Symbol | CXCL1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction