missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Clusterin Protein
A cDNA sequence encoding the Clusterin was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11454-10ug
Additional Details : Weight : 0.01000kg
Specifications
24854 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MASMTGGQQM GRDPNSSSPF YFWMNGDRID SLLESDRQQS QVLDAMQDSF TRASGIIDTL FQDRFFTHEPQDIHHFSPMG FPHKRPHLLY PKSRLVRSLM PLSHYGPLSF HNMFQPFFDM IHQAQQAMDV QLHSPALQFPDVDFLKEGED DRTVCKEIRH NSTGCLKMKG QCEKCQEILS VDCSTNNPAQ ANLRQELNDS LQVAERLTQQYNELLHSLQS KMLNTSSLLE QALEHHHHHH | |
Greater than 95% as determined by SDS PAGE. |
Clusterin Protein | |
Research Use Only | |
Clu | |
Rat | |
His | |
Lyophilized from 0.5 mg/ml in 0.01M Tris pH 7.2. |