missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Rat CD27/TNFRSF7 Fc Chimera Protein

Product Code. 18344028
Change view
Click to view available options
:
50ug; Carrier Free
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18344028 50ug; Carrier Free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18344028 Supplier R&D Systems™ Supplier No. 7888CD050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to inhibit the proliferation of mouse T cells induced by CD27 Ligand and anti-CD3. The ED50 for this effect is 0.4-1.6 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_001019506
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 939
Molecular Weight (g/mol) M.W. (Observed): 60-66 kDa, reducing conditions; M.W. (theoretical): 45.1 kDa (monomer)
Name CD27/TNFRSF7 Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived rat CD27/TNFRSF7 protein Rat CD27/TNFRSF7 (Thr21-Arg183) (N-terminus) Accession # NP_001019506 IEGRMDP Mouse IgG2A (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol Cd27
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 400 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein CD27/TNFRSF7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.