missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Rat CCL2/JE/MCP-1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 3144-JE-025
Additional Details : Weight : 0.00970kg
Description
The Recombinant Rat CCL2/JE/MCP-1 Protein is derived from NS0. The Recombinant Rat CCL2/JE/MCP-1 Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose and with BSA as a carrier protein. | |
14 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
24770 | |
CCL2/JE/MCP-1 | |
Mouse myeloma cell line,NS0-derived rat CCL2/JE/MCP-1 protein Gln24-Asn148 | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |