missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Cathepsin K Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Cathepsin K, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5892-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_113748.1 | |
Cathepsin K Protein | |
Research Use Only | |
Ctsk | |
Recombinant Protein | |
E. coli | |
VPDSIDYRKK GYVTPVKNQG QCGSCWAFSS AGALEGQLKK KTGKLLALSP QNLVDCVSEN YGCGGGYMTT AFQYVQQNGG IDSEDAYPYV GQDESCMYNA TAKAAKCRGY REIPVGNEKA LKRAVARVGP VSVSIDASLT SFQFYSRGVY YDENCDRDNV NHAVLVVGYG TQKGNKYWII KNSWGESWGN KGYVLLARNK NNACGITNLA SFPKM | |
Greater than 90% as determined by SDS-PAGE. |
29175 | |
50 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |