missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Carboxypeptidase B Protein
Click to view available options
Quantity:
5 mg
15 mg
50 mg
Unit Size:
15mg
50mg
5mg
Specifications
Specifications
| Name | Carboxypeptidase B Protein |
| Quantity | 5 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
| Biological Activity | 170 units/mg protein. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Rat |
| Species | E. coli |
| Protein Tag | Untagged |
| Sequence | MLLLLALVSV ALAHASEEHF DGNRVYRVSV HGEDHVNLIQ ELANTKEIDF WKPDSATQVK PLTTVDFHVK AEDVADVENF LEENEVHYEV LISNVRNALE SQFDSHTRAS GHSYTKYNKW ETIEAWIQQV ATDNPDLVTQ SVIGTTFEGR NMYVLKIGKT RPNKPAIFID CGFHAREWIS PAFCQWFVRE AVRTYNQEIH MKQLLDELDF YVLPVVNIDG YVYTWTKDRM WRKTRSTMAG SSCLGVRPNR NFNAGWCEVG ASRSPCSETY CGPAPESEKE TKALADFIRN NLSTIKAYLT IHSYSQMMLY PYSYDYKLPE NYEELNALVK GAAKELATLH GTKYTYGPGA TTIYPAAGGS DDWSYDQGIK YSFTFELRDT GFFGFLLPES QIRQTCEETM LAVKYIANYV REHLY |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction