missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat CAMK4 / CaMKIV Protein
A DNA sequence encoding the Rattus norvegicus (Rat) CAMK4 / CaMKIV, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5752-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_036859.2 | |
CAMK4 / CaMKIV Protein | |
Research Use Only | |
Camk4 | |
Recombinant Protein | |
E. coli | |
MLKVTVPSCP SSPCSSVTSS TENLVPDYWI DGSKRDPLSD FFEVESELGR GATSIVYRCK QKGTQKPYAL KVLKKTVDKK IVRTEIGVLL RLSHPNIIKL KEIFETPTEI SLVLELVTGG ELFDRIVEKG YYSERDAADA VKQILEAVAY LHENGIVHRD LKPENLLYAT PAPDAPLKIA DFGLSKIVEH QVLMKTVCGT PGYCAPEILR GCAYGPEVDM WSVGIITYIL LCGFEPFYDE RGDQFMFRRI LNCEYYFISP WWDEVSLNAK DLVKKLIVLD PKKRLTTFQA LQHPWVTGKA ANFVHMDTAQ KKLQEFNARR KLKAAVKAVV ASSRLGSASS SHTNIQESNK ASSEAQPAQD GKDKTDPLEN KMQAGDHEAA KAAADETMKL QSEEVEEEEG VKEEEEEEEE EEETSRMVPQ EPEDRLETDD QEMKRNSEET LKSVEEEMDP KAEEEAAAVG LGVPPQQDAI LPEY | |
Greater than 90% as determined by SDS-PAGE. |
25050 | |
10 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |