Learn More
DiscoverX Recombinant Rat Beta-NGF
Product Code. 17332599
Encompass
Click to view available options
:
100 μg
Product Code. 17332599
Brand: DiscoverX 921283
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Recombinant Rat Beta-NGF is an agonist for TrkA. Provided as 100μg of dry powder alongside reconstitution buffer.
Beta-nerve growth factor (NGF) is a prototypical growth factor that functions in sympathetic and sensory neuron survival and maintenance. Mutations have been associated with hereditary sensory and autonomic neuropathy. Extracellular ligand (agonist) for the TrkA (NTRK1) and NGFR receptors. Provided as 100μg of dry powder alongside reconstitution buffer.
Specifications
Quantity 100 μg
Product Type Control Ligands