missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Rat B7-H4 Fc Chimera Protein

Product Code. 18786256
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18786256 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18786256 Supplier R&D Systems™ Supplier No. 10085B7050
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to inhibit anti-CD3 antibody induced IL-2 or IFN-gamma secretion by human T cells. The ED50 for this effect is 1-6 μg/mL.
TRUSTED_SUSTAINABILITY

Specifica

Accession Number Q501W4
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 79679
Molecular Weight (g/mol) M.W. (Observed): 74-85 kDa, reducing conditions; M.W. (theoretical): 52 kDa
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived rat B7-H4 protein Rat B7-H4 (Phe29-Gly257) (N-terminus) Accession # Q501W4 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Specific Reactivity B7-H4
Conjugate Unconjugated
Species Rat
Recombinant Recombinant
Reconstitution Reconstitute at 500 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein B7-H4
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.