missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Rat B7-H3 Fc Chimera Protein, CF
Description
Measured by its ability to inhibit Mouse CD3 epsilon Antibody induced proliferation of Mouse T cells. The ED50 for this effect is 0.06-0.54 μg/mL.
Specifications
Specifications
| Accession Number | Q7TPB4.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 80381 |
| Molecular Weight (g/mol) | M.W. (Observed): 66-76 kDa, under reducing conditions, M.W. (Theoretical): 50 kDa |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line, NS0-derived rat B7-H3 protein Rat B7-H3 (Val29-Phe244) (N-terminus) Accession # Q7TPB4.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | B7H3, B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276, CD276 antigen, CD276 molecule, Costimulatory molecule |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?