missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Rat B7-H2 Protein

Product Code. 18336377
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18336377 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18336377 Supplier R&D Systems™ Supplier No. 9938B7100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

His-tag version

Measured by its ability to co-stimulate IL-4 secretion by D10.G4.1 mouse helper T cells in the presence of anti-CD3. The ED50 for this effect is 0.3-1.5 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number XP_006256322
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 23308
Molecular Weight (g/mol) M.W. (Observed): 47-62 kDa, reducing conditions; M.W. (theoretical): 28 kDa
Quantity 100 μg
Source Mouse myeloma cell line, NS0-derived rat B7-H2 protein Glu23-Lys261, with a C-Terminal 6-His tag
Storage Requirements 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Cross Reactivity B7-H2/ICOSLG
Species Rat
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein B7-H2/ICOSLG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.