missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Rat B7-1/CD80 Fc Chimera Protein

Product Code. 18348917
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18348917 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18348917 Supplier R&D Systems™ Supplier No. 1214B7050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to induce IL-2 secretion by Jurkat human acute T cell leukemia cells. Freeman, G.J. et al. (1993) Science 262:909. The ED50 for this effect is 0.1-0.4 μg/mL in the presence of PHA.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AAB60503
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 941
Molecular Weight (g/mol) M.W. (Observed): 80-90 kDa, reducing conditions; M.W. (theoretical): 50.5 kDa (monomer)
Name B7-1/CD80 Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived rat B7-1/CD80 protein Rat B7-1 (Ile39 - Gln248) (N-terminus) Accession # AAB60503 IEGRMD Human IgG1 (Pro100 - Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol Cd80
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein B7-1/CD80
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.