missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Apolipoprotein A-V Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Apolipoprotein A-V, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5663-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001264193.1 | |
Apolipoprotein A-V Protein | |
Research Use Only | |
Apoa5 | |
Recombinant Protein | |
E. coli | |
RKSFWEYFGQ NSQGKGMMGQ QQKLAQESLK GSLEQDLYNM NNFLEKLGPL REPGKEPPRL AQDPEGIRKQ LQQELEEVST RLEPYMAAKH QQVGWNLEGL RQQLKPYTVE LMEQVGLSVQ DLQEQLRMVG KGTKAQLLGG VDEAMSLLQD MQSRVLHHTD RVKELFHPYA ERLVTGIGHH VQELHRSVAP HAVASPARLS RCVQTLSHKL TRKAKDLHTS IQRNLDQLRD ELSTFIRVST DGADNRDSLD PQALSDEVRQ RLQAFRHDTY LQIAAFTQAI DQETEEIQHQ LAPPPPSHSA FAPELGHSDS NKALSRLQSR LDDLWEDIAY GLHDQGHSQN NPEGHSG | |
Greater than 90% as determined by SDS-PAGE. |
140638 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). of Isoform 3 | |
Rat | |
GST | |
Tris-based buffer, 50% glycerol |