missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Rat Adiponectin/Acrp30 FLAG Protein

Product Code. 18329863 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18329863 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18329863 Supplier Novus Biologicals™ Supplier No. NBP318139

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Induces the phosphorylation of acetyl-CoA carboxylase (ACC) in C2C12 cells
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8K3R4
Concentration LYOPH
For Use With (Application) Bioactivity, Western Blot
Formulation Lyophilized from 30mM Tris-Cl, pH 8.0 + 100mM NaCl
Gene ID (Entrez) 9370
Quantity 50 μg
Storage Requirements Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.01 EU/μg purified protein (LAL test).
Gene Alias ACDC, ACRP30ADPN, adipocyte, C1Q and collagen domain containing, adiponectin, adiponectin, C1Q and collagen domain containing, AdipoQ, ADIPQTL1, apM-1, APM1APM-1, C1q and collagen domain-containing protein, GBP28apM1, Gelatin-binding protein
Gene Symbol ADIPOQ
Reconstitution Reconstitute with 50 ul endotoxin-free water (1mg/ml after reconstitution)
Research Category Diabetes Research, Lipid and Metabolism
Purity or Quality Grade >95% pure by SDS-PAGE
Protein Adiponectin/Acrp30
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.