missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Acetyl-CoA carboxylase 1 Protein

Product Code. 15996379
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 15996379

Brand: enQuireBio™ QP5604ye100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Rattus norvegicus (Rat) Acetyl-CoA carboxylase 1, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_071529.1
Gene ID (Entrez) 60581
Name Acetyl-CoA carboxylase 1 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Acaca
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Rat
Species Yeast
Protein Tag His
Sequence VIEKVLIANN GIAAVKCMRS IRRWSYEMFR NERAIRFVVM VTPEDLKANA EYIKMADHYV PVPGGANNNN YANVELILDI AKRIPVQAVW AGWGHASENP KLPELLLKNG IAFMGPPSQA MWALGDKIAS SIVAQTAGIP TLPWSGSGLR VDWQENDFSK RILNVPQDLY EKGYVKDVDD GLKAAEEVGY PVMIKASEGG GGKGIRKVNN ADDFPNLFRQ VQAEVPGSPI FVMRLAKQSR HLEVQILADQ YGNAISLFGR DCSVQRRHQK IIEEAPAAIA TPAVFEHMEQ CAVKLAKMVG YVSAGTVEYL YSQDGSFYFL ELNPRLQVEH PCTEMVADVN LPAAQLQIAM GIPLFRIKDI RMMYGVSPWG DAPIDFENSA HVPCPRGHVI AARITSENPD EGFKPSSGTV QELNFRSNKN VWGYFSVAAA GGLHEFADSQ FGHCFSWGEN REEAISNMVV ALKELSIRGD FRTTVEYLIK LLETESFQLN RIDTGWLDRL IA
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.