missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rabbit Prolactin Protein
A cDNA sequence encoding the Prolactin was constructed and used to recombinantly synthesize the protein.
£99.15 - £1495.00
Specifications
Name | Prolactin Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Product Type | Recombinant Protein |
Cross Reactivity | Rabbit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15945358
|
enQuireBio™
QP10850-10UG |
10 μg |
£99.15
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965358
|
enQuireBio™
QP10850-50UG |
50 μg |
£155.00
50µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955358
|
enQuireBio™
QP10850-1MG |
1 mg |
£1495.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Prolactin Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Rabbit | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MDSKWSRRTG SLLLLLVSNL LLCKSTASLP ICPSGAVNCQ VSLRDLFDRA VILSHHIHKL SSEMFNEFDK RYTQGRGFIT KAINSCHTSS LSTPEDKEQA QQIHHEDLLN MVLRVLPSWN DPLYHLVTEV RGMQEAPDAI LSKAIEIEEQ NRRLLEGMEK IVGQVHPGIK ENEIYSVWSG LPSLQMADED ARLFAFYNLL HCLRRDSHKI DNYLKLLKCR IIYDSNC | |
Greater than 98.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |