missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rabbit MFI2 / CD228 / melanotransferrin Protein
A DNA sequence encoding the Oryctolagus cuniculus (Rabbit) MFI2 / CD228 / melanotransferrin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6357-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001075461.1 | |
MFI2 / CD228 / melanotransferrin Protein | |
Research Use Only | |
Mfi2 | |
Recombinant Protein | |
E. coli | |
SVEVRWCTAS EPEQQKCEDM SQAFREAGLQ PALLCVQGTS ADHCVQLIAA HEADAITLDG GAIYEAGKEH GLKPVVGEVY DQEVGTSYYA VAVVKRSSNV TINTLRGVKS CHTGINRTVG WNVPVGYLVD SGRLSVMGCD VLKAVSEYFG GSCVPGAGET RYSESLCRLC RGDTSGEGVC DKSPLERYYD YSGAFRCLAE GAGDVAFVKH STVLENTDGR TLPSWGHMLM SRDFELLCRD GSRASVTEWQ HCHLARVPAH AVVVRADTDA GLIFRLLNEG QRLFSHEGSS FQMFSSEAYG QKNLLFKDST LELVPIATQT YEAWLGPEYL HAMKGLLCDP NRLPPYLRWC VLSTPEIQKC GDMAVAFSRQ RLKPEIQCVS AESPQHCMEQ IQAGHIDAVT LNGEDIHTAG KTYGLIPAAG ELYAADDRSN SYFVVAVVKR DSAYAFTVDE LRGKRSCHPG FGSPAGWDVP VGALIHWGYI RPRNCDVLTA VGQFFNASCV PVNNPKKYPS SLCALCVGDE QGRNKCTGNS QERYYGDSGA FRCLVEGAGD VAFVKHTTIF DNTNGHNPEP WAAHLRSQDY ELLCPNGARA EAHQFAACNL AQIPSHAVMV RPDTNIFTVY GLLDKAQDLF GDDHNKNGFK MFDSSSYHGR DLLFKDATVR AVPVGERTTY QDWLGPDYVA ALEGMQSQRC S | |
Greater than 90% as determined by SDS-PAGE. |
100008603 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rabbit | |
His-SUMO | |
Tris-based buffer, 50% glycerol |