missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rabbit IL6 / Interleukin-6 Protein
A DNA sequence encoding the Oryctolagus cuniculus (Rabbit) IL6 / Interleukin-6 Protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9889-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001075533.1 | |
IL6 / Interleukin-6 Protein | |
Research Use Only | |
IL6 | |
Recombinant Protein | |
Yeast | |
FPTSAPVRED SNTKASPDKT LTPPGRTIES IRSILETIKE LRKEMCDHDV NCMNRKEALA EVNLHLPRLI EEDGCFPPAV NNETCLLRIT SGLMEFRMYL EHLQAKFRSD EENTRVSMVL KNIQHLIKTL RPKVKNLNEE ATLKPAVAVS LMENLQQKNQ WLKTTTIHFI LRGLTNFLEF TLRAVDLMEC GCPCLRNFMG SASHGQNTPS CPLDT | |
Greater than 90% as determined by SDS-PAGE. |
100008733 | |
10 μg | |
Not determined. | |
Full Length | |
Rabbit | |
His | |
Tris-based buffer, 50% glycerol |