missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rabbit CD40L / CD154 / TNFSF5 Protein
A DNA sequence encoding the Oryctolagus cuniculus (Rabbit) CD40L / CD154 / TNFSF5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5787-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001243710.1 | |
CD40L / CD154 / TNFSF5 Protein | |
Research Use Only | |
CD40LG | |
Recombinant Protein | |
E. coli | |
MQKGDQDPQI AAHLISEASS KSSSVLQWAK KGYYTMSNTL VTLENGKQLK VKRQGFYYIY AQVTFCSNQE PSSQAPFIAS LCLKSSGGSE RILLRAANAR SSSKTCEQQS IHLGGVFELQ ADASVFVNVT DASQVNHGTG FTSFGLLKL | |
Greater than 90% as determined by SDS-PAGE. |
100358388 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rabbit | |
His | |
Tris-based buffer, 50% glycerol |