missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pseudomonas Thiol:disulfide interchange protein DsbA Protein
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
Specifications
Specifications
| Accession Number | NP_790191.1 |
| Gene ID (Entrez) | 1181950 |
| Name | Pseudomonas Thiol:disulfide interchange protein DsbA Protein |
| Quantity | 50 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not determined. |
| Gene Symbol | dsbA |
| Protein Length | Full Length |
| Product Type | Recombinant Protein |
| Cross Reactivity | Pseudomonas |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction