missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pseudomonas Carboxypeptidase G2 Protein
A DNA sequence encoding the Pseudomonas sp. (strain RS-16) Carboxypeptidase G2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7364-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
Pseudomonas Carboxypeptidase G2 Protein | |
Research Use Only | |
cpg2 | |
Recombinant Protein | |
E. coli | |
ALAQKRDNVL FQAATDEQPA VIKTLEKLVN IETGTGDAEG IAAAGNFLEA ELKNLGFTVT RSKSAGLVVG DNIVGKIKGR GGKNLLLMSH MDTVYLKGIL AKAPFRVEGD KAYGPGIADD KGGNAVILHT LKLLKEYGVR DYGTITVLFN TDEEKGSFGS RDLIQEEAKL ADYVLSFEPT SAGDEKLSLG TSGIAYVQVN ITGKASHAGA APELGVNALV EASDLVLRTM NIDDKAKNLR FNWTIAKAGN VSNIIPASAT LNADVRYARN EDFDAAMKTL EERAQQKKLP EADVKVIVTR GRPAFNAGEG GKKLVDKAVA YYKEAGGTLG VEERTGGGTD AAYAALSGKP VIESLGLPGF GYHSDKAEYV DISAIPRRLY MAARLIMDLG AGK | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Full Length | |
Pseudomonas | |
GST | |
Tris-based buffer, 50% glycerol |