missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Protein A Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
Description
Protein A is a genetically engineered protein containing 5 IgG-binding regions of protein A. Cell wall binding region, cell membrane binding region and albumin binding region have been removed from the recombinant Protein A to ensure the maximum specific IgG binding. The recombinant Protein A is ideal for purification of polyclonal or monoclonal IgG antibodies. Protein A binds to various human, mouse and rat IgG subclasses. It also binds to total IgG from rabbit, guinea pig, pig, dog and cat.
Specifications
Specifications
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity, HPLC |
| Formulation | Lyophilized from additive free solution. |
| Gene ID (Entrez) | 3919448 |
| Molecular Weight (g/mol) | 33.4 kDa |
| Quantity | 100 mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/ug of Protein A as determined by LAL method. |
| Gene Alias | Immunoglobulin G binding protein A, SPA, Staphylococcal protein A |
| Reconstitution | Dissolve in distilled water or saline. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction