missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Protein A is a genetically engineered protein containing 5 IgG-binding regions of protein A. The recombinant Protein A is ideal for purification of polyclonal or monoclonal IgG antibodies. Protein A binds to various human, mouse and rat IgG subclasses (e.g., human IgG1, IgG2, IgG4; mouse IgG2a, IgG2b, IgG3; rat IgG2c). It also binds to total IgG from rabbit, guinea pig, pig, dog and cat.
Specifications
Specifications
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from additive free solution. |
| Gene ID (Entrez) | 3919448 |
| Molecular Weight (g/mol) | 46.6 kDa |
| Quantity | 10 mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/ug of Protein A as determined by LAL method. |
| Gene Alias | Immunoglobulin G binding protein A, SPA, Staphylococcal protein A |
| Reconstitution | Dissolve in distilled water or saline. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?