missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Protein A/G Protein

Product Code. p-200055411 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 mg
Unit Size:
100mg
1mg
This item is not returnable. View return policy

Product Code. 18420372

Brand: Novus Biologicals™ NBP234968100mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Protein A/G is a genetically engineered protein containing 5 IgG-binding regions of protein A and 2 of protein G. Cell wall binding region, cell membrane binding region and albumin binding region have been removed from Protein A/G to ensure the maximum specific IgG binding. Protein A/G is ideal for purification of polyclonal or monoclonal IgG antibodies. It binds to various human, mouse and rat IgG subclasses. It also binds to total IgG from cow, goat, sheep, house, rabbit, guinea pig, pig, dog and cat.
TRUSTED_SUSTAINABILITY

Specifications

Concentration LYOPH
For Use With (Application) PAGE, Bioactivity
Formulation Lyophilized from additive free solution.
Molecular Weight (g/mol) 47.7 kDa
Quantity 100 mg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 0.1 EU/ug of Protein A/G as determined by LAL method.
Reconstitution Dissolve in distilled water or saline.
Research Category Biologically Active Proteins
Purity or Quality Grade >97% pure by SDS-PAGE and HPLC
Protein Protein A/G
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.