missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Porcine IL-5 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 3137-PL-010/CF

Additional Details : Weight : 0.00970kg

 View more versions of this product

Product Code. 18146250

  • £292.65 / 10µg
Estimated Shipment: 03-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Porcine IL-5 Protein is derived from NS0. The Recombinant Porcine IL-5 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
14 kDa (monomer)
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
397409
IL-5
Mouse myeloma cell line,NS0-derived porcine IL-5 protein Ile20-Ser134,with a C-terminal 6-His tag
Disulfide-linked homodimer
Recombinant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers