missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Porcine IL-18/IL-1F4 Protein
Description
Measured by its ability to induce IFN-gamma secretion by KG-1 human acute myelogenous leukemia cells in the presence of TNF-alpha . The ED50 for this effect is 10-40 ng/mL in the presence of 20 ng/mL rhTNF-alpha .
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS and DTT with Trehalose. |
| Gene ID (Entrez) | 397057 |
| Molecular Weight (g/mol) | 18.0 kDa |
| Name | IL-18/IL-1F4 |
| Quantity | 25 μg |
| Source | E. coli-derived porcine IL-18/IL-1F4 protein Tyr36-Asn192 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?