missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Plasmodium Vivax Circumsporozoite (CSP) Protein

Product Code. 18673237 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
This item is not returnable. View return policy

Product Code. 18673237

Brand: Novus Biologicals™ NBP262522

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/mL
For Use With (Application) ELISA
Formulation PBS
Quantity 0.1 mg
Storage Requirements Store at -80°C. Avoid freeze/thaw cycles.
Gene Alias P. vivax, Plasmodium vivax circumsporozoite protein
Protein Plasmodium Vivax Circumsporozoite (CSP)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.