missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Plant Apium graveolens 1.0101 His (C-Term) Protein
Specifications
Specifications
| For Use With (Application) | ELISA, Flow Cytometry, SDS-PAGE |
| Formulation | Lyophilized from a 0.22 um filtered solution in Hepes buffer (20 mM Hepes, 300 mM NaCl, 2% glycerol, 0.01% Tween 20, pH 7.4) |
| Molecular Weight (g/mol) | M.W. Theoretical: 17.9 kDa |
| Quantity | 0.1 mg |
| Storage Requirements | Store at -20 to -70C. Avoid freeze-thaw cycles. |
| Gene Alias | rApi g 1 |
| Reconstitution | Reconstitute with sterile deionized water with or without azide. |
| Purity or Quality Grade | >95%, by HPLC |
| Protein | Apium graveolens 1.0101 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?