missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | Lyophilized from a 0.2 um filtered solution in Storage buffer (100 mM Tris, 150 mM NaCl, 0.01% Tween 20, pH 8.0). |
| Quantity | 0.1 mg |
| Storage Requirements | Store at -20 to -70C. Avoid freeze-thaw cycles. |
| Gene Alias | Ambrosia artemisiifolia 1.0101, Antigen E |
| Reconstitution | Reconstitute in sterile deionized water. Use reconstituted product immediately or aliquot for further storage at -20C to -80C. |
| Purity or Quality Grade | Multi-step |
| Protein | Amb a 1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?