missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant P. falciparum HSP90 Protein

Product Code. 18370235
Change view
Click to view available options
Quantity:
100 μg
200 μg
50 μg
Unit Size:
100µg
200µg
50µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18370235 100 μg 100µg
18300472 50 μg 50µg
18334077 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18370235 Supplier Novus Biologicals™ Supplier No. NBP318321100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) SDS-PAGE, Western Blot
Formulation 50mM Tris/HCl pH7.5, 300mM NaCl, 10% glycerol
Gene ID (Entrez) 3320
Molecular Weight (g/mol) M.W. Theoretical: 21.4 kDa
Quantity 100 μg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias FLJ31884, Heat shock 86 kDa, heat shock 90kD protein 1, alpha, heat shock 90kD protein 1, alpha-like 4, heat shock 90kD protein, alpha-like 4, heat shock 90kDa protein 1, alpha, heat shock protein 90kDa alpha (cytosolic), class A member 1, heat shock protein HSP 90-alpha, Hsp89, Hsp90, HSP90AHSP86, HSP90N, HSPC1HSP 86, HSPCAHSP89A, HSPCAL1, HSPCAL4, HSPN, LAP2, Renal carcinoma antigen NY-REN-38
Gene Symbol HSP90AA1
Research Category Cancer, Hypoxia, Membrane Trafficking and Chaperones
Purity or Quality Grade >85%
Protein HSP90
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.