missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant P. aeruginosa PA-I galactophilic lectin Protein

A DNA sequence encoding the Pseudomonas aeruginosa (strain ATCC 15692 / PAO1 / 1C / PRS 101 / LMG 12228) PA-I galactophilic lectin, was expressed in the hosts and tags indicated. option, ab

Brand:  enQuireBio™ QP6995-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16076940

  • £320.00 / 10µg
Estimated Shipment: 15-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_251260.1
P. aeruginosa PA-I galactophilic lectin Protein
Research Use Only
lecA
Recombinant Protein
E. coli
AWKGEVLANN EAGQVTSIIY NPGDVITIVA AGWASYGPTQ KWGPQGDREH PDQGLICHDA FCGALVMKIG NSGTIPVNTG LFRWVAPNNV QGAITLIYND VPGTYGNNSG SFSVNIGKDQ S
Greater than 90% as determined by SDS-PAGE.
882335
10 μg
Not determined.
Full Length
P. aeruginosa
His
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers