missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Ovine GH Protein
A cDNA sequence encoding the GH was constructed and used to recombinantly synthesize the protein.
£99.15 - £1165.00
Specifications
Name | GH Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | Ovine placental Grwoth Hormone is fully biologically active when compared to World Health Organization (WHO) reference standard using in vitro bioassay in PDF-P1 3B9 cells stably transfected with rabbit GH receptors. It is also capable of forming a 1:2 complex with the recombinant ovine growth hormone receptor extracellular domain (ECD). |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15954478
|
enQuireBio™
QP10634-20UG |
20 μg |
£99.15
20µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15934478
|
enQuireBio™
QP10634-100UG |
100 μg |
£155.00
100µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15944478
|
enQuireBio™
QP10634-1MG |
1 mg |
£1165.00
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
GH Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3 adjusted to pH 9. |
Research Use Only | |
Ovine placental Grwoth Hormone is fully biologically active when compared to World Health Organization (WHO) reference standard using in vitro bioassay in PDF-P1 3B9 cells stably transfected with rabbit GH receptors. It is also capable of forming a 1:2 complex with the recombinant ovine growth hormone receptor extracellular domain (ECD). | |
Ovine | |
Untagged | |
Greater than 98.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |