missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other TBEV NS3 Protein

A cDNA sequence encoding the TBEV NS3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13702-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15944429

  • £173.00 / 100µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TBEV NS3 Protein
Research Use Only
Recombinant Protein
E. coli
20mM MES pH 6.5, 8M urea, 200mM NaCl and 0.05% Tween-20.
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Encephalitis protein is >95% pure as determined by 10% PAGE (coomassie staining).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers