missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other T. pallidum TmpA Protein
A cDNA sequence encoding the T. pallidum TmpA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13677-1mg
Additional Details : Weight : 0.01000kg
Specifications
T. pallidum TmpA Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
8M urea, 20mM Tris-HCl pH 8 and 10mM B-ME. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Treponema Pallidum protein is >90% pure as determined by 10% PAGE (coomassie staining). |