missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other T. pallidum p41 Mosaic Protein

Product Code. 15994339
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15994339 500 μg 500µg
15974339 100 μg 100µg
15984339 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15994339 Supplier enQuireBio™ Supplier No. QP13674500ug
  • ONLINE EXCLUSIVE PRICE
    £733.95
    £697.83 / 500µg
    Save £36.12
    4% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A cDNA sequence encoding the T. pallidum p41 Mosaic was constructed and used to recombinantly synthesize the protein.

Specifications

Name T. pallidum p41 Mosaic Protein
Quantity 500 μg
Regulatory Status RUO
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Other
Species E. coli
Protein Tag GST
Buffer 25mM Tris-HCl pH 8.0, 60mM NaCl and 50% glycerol.
Purity or Quality Grade Treponema Pallidum protein is >90% pure as determined by 10% PAGE (coomassie staining).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.