missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other T. pallidum p41 Mosaic Protein
Specifications
Specifications
| Name | T. pallidum p41 Mosaic Protein |
| Quantity | 100 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Other |
| Species | E. coli |
| Protein Tag | GST |
| Buffer | 25mM Tris-HCl pH 8.0, 60mM NaCl and 50% glycerol. |
| Purity or Quality Grade | Treponema Pallidum protein is >90% pure as determined by 10% PAGE (coomassie staining). |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?